siRNA Details
| Gene Symbol: Pid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110493.1 | NP_001103963.1 | 3 | 145 | ||
| GenBank mRNA | AY325140.1 | - | 3 | 145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pid1| Gene Symbol: Pid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110493.1 | NP_001103963.1 | 3 | 145 | ||
| GenBank mRNA | AY325140.1 | - | 3 | 145 | ||