siRNA Details
| Gene Symbol: Cr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105989.2 | NP_001099459.2 | 11 | 1993 | ||
| XM_006250476.2 | XP_006250538.1 | 15 | 2608 | |||
| XM_006250477.2 | XP_006250539.1 | 11 | 2062 | |||
| XM_008769847.1 | XP_008768069.1 | 18 | 3193 | |||
| XM_008769848.1 | XP_008768070.1 | 11 | 2479 | |||

Cr2
Genomic Map