siRNA Details
| Gene Symbol: Grhpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113754.1 | NP_001107226.1 | 7, 8 | 800 | ||
| GenBank mRNA | BC158680.1 | - | 7, 8 | 784 | 8373346 | |
| FQ210026.1 | - | 7, 8 | 771 | |||
| FQ210631.1 | - | 7, 8 | 747 | |||

Grhpr
Genomic Map