siRNA Details
| Gene Symbol: Grhpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113754.1 | NP_001107226.1 | 6 | 572 | ||
| GenBank mRNA | BC158680.1 | - | 6 | 556 | 8373346 | |
| FQ210026.1 | - | 6 | 543 | |||
| FQ210631.1 | - | 6 | 519 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grhpr| Gene Symbol: Grhpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113754.1 | NP_001107226.1 | 6 | 572 | ||
| GenBank mRNA | BC158680.1 | - | 6 | 556 | 8373346 | |
| FQ210026.1 | - | 6 | 543 | |||
| FQ210631.1 | - | 6 | 519 | |||