siRNA Details
| Gene Symbol: Sprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031845.1 | NP_001027015.1 | 1 | 141 | ||
| XM_006230573.2 | XP_006230635.1 | 2 | 418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprn| Gene Symbol: Sprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031845.1 | NP_001027015.1 | 1 | 141 | ||
| XM_006230573.2 | XP_006230635.1 | 2 | 418 | |||