siRNA Details
| Gene Symbol: Ccnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059113.1 | - | 6 | 846 | 6922392 | |
| EV765751.1 | - | 2 | 251 | |||
| FN800627.1 | - | 4 | 499 | |||
| L11995.1 | - | 6 | 838 | |||
| X60768.1 | - | 6 | 776 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnb1| Gene Symbol: Ccnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059113.1 | - | 6 | 846 | 6922392 | |
| EV765751.1 | - | 2 | 251 | |||
| FN800627.1 | - | 4 | 499 | |||
| L11995.1 | - | 6 | 838 | |||
| X60768.1 | - | 6 | 776 | |||