siRNA Details
| Gene Symbol: Arhgap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170575.1 | NP_001164046.1 | 14 | 1954 | ||
| XM_006227680.2 | XP_006227742.1 | 14 | 2740 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap18| Gene Symbol: Arhgap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170575.1 | NP_001164046.1 | 14 | 1954 | ||
| XM_006227680.2 | XP_006227742.1 | 14 | 2740 | |||