siRNA Details
| Gene Symbol: Slc39a4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101021.1 | NP_001094491.1 | 1110 | |||
| GenBank mRNA | DQ256461.1 | - | 1110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc39a4l| Gene Symbol: Slc39a4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101021.1 | NP_001094491.1 | 1110 | |||
| GenBank mRNA | DQ256461.1 | - | 1110 | |||