siRNA Details
| Gene Symbol: Usp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107827.3 | NP_001101297.1 | 21 | 2413 | ||
| XM_006233916.2 | XP_006233978.1 | 21 | 3548 | |||
| XM_008761666.1 | XP_008759888.1 | 19 | 2522 | |||
| XM_008761667.1 | XP_008759889.1 | 19 | 2480 | |||

Usp20
Genomic Map