siRNA Details
| Gene Symbol: DPY19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015283.1 | NP_056098.1 | 8 | 817 | ||
| XM_011515245.2 | XP_011513547.1 | 8 | 966 | |||
| XM_011515246.2 | XP_011513548.1 | 8 | 966 | |||
| GenBank mRNA | AB020684.1 | - | 5 | 388 | ||
| DQ287932.1 | - | 8 | 817 | |||

DPY19L1
Genomic Map