siRNA Details
| Gene Symbol: Tmsb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021261.2 | NP_067084.1 | 3 | 414 | ||
| XM_006236692.2 | XP_006236754.1 | 3 | 465 | |||
| GenBank mRNA | BC126092.1 | - | 3 | 383 | ||
| FQ221466.1 | - | 3 | 412 | |||
| FQ223015.1 | - | 3 | 412 | |||
| FQ224120.1 | - | 3 | 414 | |||
| FQ224544.1 | - | 3 | 412 | |||
| FQ229044.1 | - | 3 | 413 | |||
| M17698.1 | - | 3 | 392 | |||
| M58404.1 | - | 2 | 487 | |||
| M58405.1 | - | 3 | 394 | |||

Tmsb10
Genomic Map