siRNA Details
| Gene Symbol: Lman2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115024.2 | NP_001108496.1 | 5 | 584 | ||
| GenBank mRNA | BC158808.1 | - | 5 | 584 | 7324877 | |
| FQ215124.1 | - | 5 | 766 | |||
| FQ227127.1 | - | 5 | 468 | |||
| FQ230263.1 | - | 5 | 582 | |||

Lman2
Genomic Map