siRNA Details
| Gene Symbol: Usp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127638.1 | NP_001121110.1 | 2 | 387 | ||
| XM_008769764.1 | XP_008767986.1 | 2 | 530 | |||
| GenBank mRNA | BC166443.1 | - | 2 | 387 | 8363035 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp21| Gene Symbol: Usp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127638.1 | NP_001121110.1 | 2 | 387 | ||
| XM_008769764.1 | XP_008767986.1 | 2 | 530 | |||
| GenBank mRNA | BC166443.1 | - | 2 | 387 | 8363035 | |