siRNA Details
| Gene Symbol: WDR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015285.2 | NP_056100.2 | 11 | 1388 | ||
| NM_052834.2 | NP_443066.2 | 11 | 1388 | |||
| XM_006722431.2 | XP_006722494.1 | 11 | 1320 | |||
| XM_011525888.1 | XP_011524190.1 | 11 | 1430 | |||
| XM_017025682.1 | XP_016881171.1 | 11 | 1293 | |||
| GenBank mRNA | AB011113.2 | - | 11 | 1351 | ||
| AF188125.1 | - | 10 | 1177 | |||
| AK057141.1 | - | 11 | 1411 | |||
| AY099325.1 | - | 10 | 1177 | |||
| BC150282.1 | - | 11 | 1351 | 9007268 | ||

WDR7
Genomic Map