siRNA Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058209.2 | NP_478116.2 | 440 | |||
| GenBank mRNA | AF072439.1 | - | 465 | |||
| BC166844.1 | - | 414 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp37| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058209.2 | NP_478116.2 | 440 | |||
| GenBank mRNA | AF072439.1 | - | 465 | |||
| BC166844.1 | - | 414 | ||||