siRNA Details
| Gene Symbol: Epc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751704.2 | XP_003751752.1 | 1 | 256 | ||
| GenBank mRNA | BC098858.1 | - | 1 | 270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epc1| Gene Symbol: Epc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751704.2 | XP_003751752.1 | 1 | 256 | ||
| GenBank mRNA | BC098858.1 | - | 1 | 270 | ||