siRNA Details
| Gene Symbol: Nr1i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270838.1 | NP_001257767.1 | 8 | 1053 | ||
| NM_001270839.1 | NP_001257768.1 | 8 | 1049 | |||
| NM_001270840.1 | NP_001257769.1 | 8 | 1050 | |||
| NM_022941.4 | NP_075230.1 | 8 | 1053 | |||
| XM_006250261.2 | XP_006250323.1 | 8 | 1159 | |||
| XM_008769759.1 | XP_008767981.1 | 8 | 1161 | |||
| GenBank mRNA | AF133094.1 | - | 7 | 904 | ||
| BC089763.1 | - | 8 | 1050 | 7324318 | ||

Nr1i3
Genomic Map