siRNA Details
| Gene Symbol: Ccnjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037773.4 | NP_001032862.2 | 3, 4 | 572 | ||
| XM_006246133.2 | XP_006246195.1 | 4, 5 | 1258 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnjl| Gene Symbol: Ccnjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037773.4 | NP_001032862.2 | 3, 4 | 572 | ||
| XM_006246133.2 | XP_006246195.1 | 4, 5 | 1258 | |||