siRNA Details
| Gene Symbol: RGD1561676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107438.1 | NP_001100908.1 | 2 | 244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561676| Gene Symbol: RGD1561676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107438.1 | NP_001100908.1 | 2 | 244 | ||