siRNA Details
| Gene Symbol: Pamr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107755.1 | NP_001101225.1 | 4 | 475 | ||
| XM_008762061.1 | XP_008760283.1 | 4 | 592 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pamr1| Gene Symbol: Pamr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107755.1 | NP_001101225.1 | 4 | 475 | ||
| XM_008762061.1 | XP_008760283.1 | 4 | 592 | |||