siRNA Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145670.4 | NP_663703.3 | 8 | 1252 | ||
| XM_008762466.1 | XP_008760688.1 | 9 | 1387 | |||
| GenBank mRNA | AB180269.1 | - | 7 | 895 | ||
| BC103480.1 | - | 7 | 1150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcas1| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145670.4 | NP_663703.3 | 8 | 1252 | ||
| XM_008762466.1 | XP_008760688.1 | 9 | 1387 | |||
| GenBank mRNA | AB180269.1 | - | 7 | 895 | ||
| BC103480.1 | - | 7 | 1150 | |||