siRNA Details
| Gene Symbol: Ap3b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748891.3 | XP_003748939.1 | 23 | 3132 | ||
| XM_006229468.2 | XP_006229530.1 | 23 | 3166 | |||
| XM_006229469.2 | XP_006229531.1 | 23 | 3137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap3b2| Gene Symbol: Ap3b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748891.3 | XP_003748939.1 | 23 | 3132 | ||
| XM_006229468.2 | XP_006229530.1 | 23 | 3166 | |||
| XM_006229469.2 | XP_006229531.1 | 23 | 3137 | |||