siRNA Details
Gene Symbol: Nkx2-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127653.1 | NP_001121125.1 | 2 | 105 | ||
GenBank mRNA | BC160896.1 | - | 2 | 105 | 7453070 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nkx2-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127653.1 | NP_001121125.1 | 2 | 105 | ||
GenBank mRNA | BC160896.1 | - | 2 | 105 | 7453070 |