siRNA Details
| Gene Symbol: Tmem151a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107570.2 | NP_001101040.1 | 1 | 1034 | ||
| XM_003749041.3 | XP_003749089.1 | 2 | 1685 | |||
| GenBank mRNA | BC168914.1 | - | 1 | 1034 | 7313547 | |
| Gene Symbol: LOC103689952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230793.2 | XP_006230855.1 | 1666 | |||

Tmem151a
Genomic Map