siRNA Details
| Gene Symbol: Olr995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000938.1 | NP_001000938.1 | 235 | |||
| Gene Symbol: Olr886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001382.1 | NP_001001382.1 | 1 | 223 | ||

Olr995
Genomic Map