siRNA Details
| Gene Symbol: B3gat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128184.1 | NP_001121656.1 | 3 | 749 | ||
| GenBank mRNA | BC166510.1 | - | 3 | 627 | 7127430 | |
| BC168710.1 | - | 3 | 730 | 8364370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3gat3| Gene Symbol: B3gat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128184.1 | NP_001121656.1 | 3 | 749 | ||
| GenBank mRNA | BC166510.1 | - | 3 | 627 | 7127430 | |
| BC168710.1 | - | 3 | 730 | 8364370 | ||