siRNA Details
| Gene Symbol: Emp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030847.1 | NP_110474.1 | 3 | 337 | ||
| XM_006229158.2 | XP_006229220.1 | 3 | 401 | |||
| XM_008759418.1 | XP_008757640.1 | 3 | 279 | |||
| GenBank mRNA | BC088131.1 | - | 3 | 333 | 7365466 | |
| FQ233038.1 | - | 3 | 324 | |||
| Y10889.2 | - | 3 | 337 | |||

Emp3
Genomic Map