siRNA Details
| Gene Symbol: Selm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115013.1 | NP_001108485.1 | 2 | 203 | ||
| GenBank mRNA | CB607044.1 | - | 2 | 175 | ||
| FQ223498.1 | - | 2 | 158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Selm| Gene Symbol: Selm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115013.1 | NP_001108485.1 | 2 | 203 | ||
| GenBank mRNA | CB607044.1 | - | 2 | 175 | ||
| FQ223498.1 | - | 2 | 158 | |||