siRNA Details
| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107245.2 | NP_001100715.2 | 4 | 413 | ||
| GenBank mRNA | BC167000.1 | - | 4 | 413 | 9031673 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcgf3| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107245.2 | NP_001100715.2 | 4 | 413 | ||
| GenBank mRNA | BC167000.1 | - | 4 | 413 | 9031673 | |