siRNA Details
| Gene Symbol: Gys1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109615.1 | NP_001103085.1 | 6 | 1088 | ||
| GenBank mRNA | BC131849.1 | - | 6 | 1045 | 7108272 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gys1| Gene Symbol: Gys1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109615.1 | NP_001103085.1 | 6 | 1088 | ||
| GenBank mRNA | BC131849.1 | - | 6 | 1045 | 7108272 | |