siRNA Details
| Gene Symbol: Zfp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107467.2 | NP_001100937.1 | 4 | 1032 | ||
| XM_006228023.2 | XP_006228085.1 | 4 | 1397 | |||
| XM_006228024.2 | XP_006228086.1 | 4 | 1241 | |||
| XM_006228025.1 | XP_006228087.1 | 4 | 1001 | |||
| XM_006228026.2 | XP_006228088.1 | 4 | 970 | |||
| XM_008758762.1 | XP_008756984.1 | 4 | 1397 | |||
| GenBank mRNA | BC168865.1 | - | 4 | 1032 | 7134370 | |

Zfp51
Genomic Map