siRNA Details
| Gene Symbol: Syndig1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109485.1 | NP_001102955.1 | 4 | 880 | ||
| XM_006240353.2 | XP_006240415.1 | 3 | 782 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syndig1l| Gene Symbol: Syndig1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109485.1 | NP_001102955.1 | 4 | 880 | ||
| XM_006240353.2 | XP_006240415.1 | 3 | 782 | |||