siRNA Details
| Gene Symbol: Diras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108987.1 | NP_001102457.1 | 2 | 1220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Diras1| Gene Symbol: Diras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108987.1 | NP_001102457.1 | 2 | 1220 | ||