siRNA Details
| Gene Symbol: Etnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108343.1 | NP_001101813.1 | 2 | 277 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etnk2| Gene Symbol: Etnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108343.1 | NP_001101813.1 | 2 | 277 | ||