siRNA Details
| Gene Symbol: Col11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212528.1 | NP_997693.1 | 4 | 705 | ||
| XM_006255928.2 | XP_006255990.1 | 4 | 1553 | |||
| XM_006255929.2 | XP_006255991.1 | 4 | 1553 | |||
| XM_008772720.1 | XP_008770942.1 | 4 | 1554 | |||
| XM_008772721.1 | XP_008770943.1 | 4 | 1659 | |||
| Gene Symbol: LOC100911886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772788.1 | XP_008771010.1 | 4 | 433 | ||

Col11a2
Genomic Map