siRNA Details
| Gene Symbol: Col11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212528.1 | NP_997693.1 | 59 | 4416 | ||
| XM_006255928.2 | XP_006255990.1 | 63 | 5621 | |||
| XM_006255929.2 | XP_006255991.1 | 60 | 5300 | |||
| XM_008772720.1 | XP_008770942.1 | 65 | 5745 | |||

Col11a2
Genomic Map