siRNA Details
| Gene Symbol: Ppp1r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138521.1 | NP_612530.1 | 4 | 617 | ||
| GenBank mRNA | AY601872.1 | - | 4 | 617 | ||
| BC078954.1 | - | 4 | 547 | 7110316 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r1b| Gene Symbol: Ppp1r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138521.1 | NP_612530.1 | 4 | 617 | ||
| GenBank mRNA | AY601872.1 | - | 4 | 617 | ||
| BC078954.1 | - | 4 | 547 | 7110316 | ||