siRNA Details
| Gene Symbol: Tesb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113183.1 | NP_001106654.1 | 7, 8 | 380 | ||
| XM_008772754.1 | XP_008770976.1 | 18, 19 | 995 | |||
| XM_008772755.1 | XP_008770977.1 | 18, 19 | 1097 | |||
| XM_008772756.1 | XP_008770978.1 | 17, 18 | 872 | |||
| XM_008772757.1 | XP_008770979.1 | 18, 19 | 995 | |||
| XM_008772758.1 | XP_008770980.1 | 18, 19 | 995 | |||
| XM_008772759.1 | XP_008770981.1 | 15, 16 | 932 | |||
| XM_008772760.1 | XP_008770982.1 | 12, 13 | 821 | |||
| GenBank mRNA | BC083628.1 | - | 13, 14 | 550 | ||
| BC097390.1 | - | 7, 8 | 380 | 7453125 | ||

Tesb
Genomic Map