siRNA Details
| Gene Symbol: Klc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749042.3 | XP_003749090.1 | 3 | 650 | ||
| XM_003749043.3 | XP_003749091.1 | 3 | 613 | |||
| XM_006230934.1 | XP_006230996.1 | 3 | 726 | |||
| XM_006230935.2 | XP_006230997.1 | 3 | 596 | |||
| XM_006230936.1 | XP_006230998.1 | 3 | 597 | |||
| XM_006230937.2 | XP_006230999.1 | 3 | 545 | |||
| XM_008760196.1 | XP_008758418.1 | 2 | 1034 | |||
| GenBank mRNA | BC166555.1 | - | 3 | 618 | 7311621 | |

Klc2
Genomic Map