siRNA Details
| Gene Symbol: Rere | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053885.2 | NP_446337.2 | 22, 23 | 5220 | ||
| XM_006239438.2 | XP_006239500.1 | 22, 23 | 5314 | |||
| XM_006239441.2 | XP_006239503.1 | 16, 17 | 7224 | |||
| XM_008764198.1 | XP_008762420.1 | 22, 23 | 5317 | |||
| XM_008764199.1 | XP_008762421.1 | 22, 23 | 5734 | |||
| XM_008764200.1 | XP_008762422.1 | 22, 23 | 5290 | |||
| XM_008764201.1 | XP_008762423.1 | 21, 22 | 5192 | |||
| XM_008764202.1 | XP_008762424.1 | 21, 22 | 4758 | |||
| XM_008764203.1 | XP_008762425.1 | 20, 21 | 6885 | |||
| GenBank mRNA | CA511326.1 | - | 3, 4 | 374 | ||
| U44091.1 | - | 21, 22 | 6029 | |||

Rere
Genomic Map