siRNA Details
| Gene Symbol: Rere | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053885.2 | NP_446337.2 | 2 | 762 | ||
| XM_006239438.2 | XP_006239500.1 | 2 | 835 | |||
| XM_008764198.1 | XP_008762420.1 | 2 | 835 | |||
| XM_008764199.1 | XP_008762421.1 | 2 | 1252 | |||
| XM_008764200.1 | XP_008762422.1 | 2 | 835 | |||
| XM_008764201.1 | XP_008762423.1 | 2 | 836 | |||

Rere
Genomic Map