siRNA Details
| Gene Symbol: Fam101a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109547.1 | NP_001103017.1 | 3 | 492 | ||
| XM_008769261.1 | XP_008767483.1 | 3 | 242 | |||
| XM_008769262.1 | XP_008767484.1 | 3 | 828 | |||
| GenBank mRNA | FQ235329.1 | - | 3 | 281 | ||

Fam101a
Genomic Map