siRNA Details
| Gene Symbol: Rab8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053998.2 | NP_446450.2 | 4 | 313 | ||
| GenBank mRNA | BC071176.1 | - | 4 | 306 | ||
| BC087584.1 | - | 3 | 144 | |||
| BC105863.1 | - | 4 | 313 | 7457017 | ||
| JN019799.1 | - | 4 | 308 | |||

Rab8a
Genomic Map