siRNA Details
| Gene Symbol: Tmprss9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047100.2 | NP_001040565.2 | 5 | 503 | ||
| XM_006240931.1 | XP_006240993.1 | 7 | 1107 | |||
| GenBank mRNA | AB109392.1 | - | 6 | 617 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss9| Gene Symbol: Tmprss9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047100.2 | NP_001040565.2 | 5 | 503 | ||
| XM_006240931.1 | XP_006240993.1 | 7 | 1107 | |||
| GenBank mRNA | AB109392.1 | - | 6 | 617 | ||