siRNA Details
| Gene Symbol: Olr1366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000980.1 | NP_001000980.1 | 1 | 902 | ||
| XM_008767521.1 | XP_008765743.1 | 2 | 1052 | |||
| XM_008767522.1 | XP_008765744.1 | 2 | 991 | |||
| XM_008767523.1 | XP_008765745.1 | 5 | 1073 | |||
| GenBank mRNA | BC097272.1 | - | 5 | 1043 | ||

Olr1366
Genomic Map