siRNA Details
| Gene Symbol: U2SURP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080415.1 | NP_001073884.1 | 16 | 1576 | ||
| NM_001320219.1 | NP_001307148.1 | 16 | 1573 | |||
| NM_001320220.1 | NP_001307149.1 | 15 | 1532 | |||
| NM_001320222.1 | NP_001307151.1 | 17 | 1641 | |||
| XM_005247248.4 | XP_005247305.1 | 16 | 1582 | |||
| XM_017006037.1 | XP_016861526.1 | 12 | 1184 | |||
| XM_017006038.1 | XP_016861527.1 | 15 | 1468 | |||
| XM_017006039.1 | XP_016861528.1 | 9 | 911 | |||
| GenBank mRNA | AB002330.1 | - | 16 | 1475 | ||
| AK057679.1 | - | 3 | 1570 | |||
| AK074327.1 | - | 9 | 840 | |||
| BC006474.1 | - | 16 | 1472 | |||
| BC111692.1 | - | 15 | 1903 | 40028600 | ||

U2SURP
Genomic Map