siRNA Details
| Gene Symbol: Wdr83os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105947.1 | NP_001099417.1 | 3 | 293 | ||
| XM_006255237.1 | XP_006255299.1 | 4 | 522 | |||
| GenBank mRNA | BC166543.1 | - | 4 | 398 | 7300989 | |
| FQ225008.1 | - | 4 | 527 | |||

Wdr83os
Genomic Map