siRNA Details
| Gene Symbol: Lmntd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127540.1 | NP_001121012.1 | 9 | 1160 | ||
| XM_006230554.2 | XP_006230616.1 | 9 | 1178 | |||
| XM_006230555.2 | XP_006230617.1 | 10 | 1239 | |||
| XM_008759991.1 | XP_008758213.1 | 10 | 1228 | |||
| XM_008759992.1 | XP_008758214.1 | 9 | 1167 | |||
| GenBank mRNA | BC162052.1 | - | 9 | 1160 | 7112915 | |

Lmntd2
Genomic Map