siRNA Details
| Gene Symbol: Gpr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023099.1 | NP_075587.1 | 1 | 834 | ||
| GenBank mRNA | AB040802.1 | - | 1 | 834 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr27| Gene Symbol: Gpr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023099.1 | NP_075587.1 | 1 | 834 | ||
| GenBank mRNA | AB040802.1 | - | 1 | 834 | ||
Set Membership: |
|