siRNA Details
| Gene Symbol: Antxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044249.2 | NP_001037714.1 | 10, 11 | 1186 | ||
| GenBank mRNA | AY754025.1 | - | 9, 10 | 617 | ||
| BC131853.1 | - | 10, 11 | 1186 | 7320366 | ||
| DQ789143.1 | - | 10, 11 | 835 | |||

Antxr1
Genomic Map